missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SPRR1A Full-length ORF (NP_005978.2, 1 a.a. - 89 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006698-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16160952

  • 545.00€ / 25µg

Estimated Shipment: 24-08-2022
to see stock.
Add to basket

Description

Description

Sequence: MNSQQQKQPCTPPPQPQQQQVKQPCQPPPQEPCIPKTKEPCHPKVPEPCHPKVPEPCQPKVPEPCQPKVPEPCPSTVTPAPAQQKTKQK
Specifications

Specifications

NP_005978.2
Liquid
6698
SPRR1A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNSQQQKQPCTPPPQPQQQQVKQPCQPPPQEPCIPKTKEPCHPKVPEPCHPKVPEPCQPKVPEPCQPKVPEPCPSTVTPAPAQQKTKQK
RUO
SPRR1A
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SPRK
SPRR1A
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers