missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SPINK2 Partial ORF (NP_066937.1, 30 a.a. - 84 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006691-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16125785

  • 508.00€ / 25µg
Estimated Shipment: 06-06-2024
to see stock.
Add to basket

Description

Description

Sequence: KYRTPNCSQYRLPGCPRHFNPVCGSDMSTYANECTLCMKIREGGHNIKIIRNGPC
Specifications

Specifications

NP_066937.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.79kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KYRTPNCSQYRLPGCPRHFNPVCGSDMSTYANECTLCMKIREGGHNIKIIRNGPC
RUO
SPINK2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6691
SPINK2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HUSI-II
SPINK2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers