missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SP110 Partial ORF (NP_004501, 271 a.a. - 380 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003431-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16159834

  • 335.00€ / 10µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

The nuclear body is a multiprotein complex that may have a role in the regulation of gene transcription. This gene is a member of the SP100/SP140 family of nuclear body proteins and encodes a leukocyte-specific nuclear body component. The protein can function as an activator of gene transcription and may serve as a nuclear hormone receptor coactivator. In addition, it has been suggested that the protein may play a role in ribosome biogenesis and in the induction of myeloid cell differentiation. Alternative splicing has been observed for this gene and three transcript variants, encoding distinct isoforms, have been identified. [provided by RefSeq]

Sequence: TPSDKKGKKRKRCIWSTPKRRHKKKSLPRGTASSRHGIQKKLKRVDQVPQKKDDSTCNSTVETRAQKARTECARKSRSEEIIDGTSEMNEGKRSQKTPSTPRRVTQGAAS
Specifications

Specifications

NP_004501
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TPSDKKGKKRKRCIWSTPKRRHKKKSLPRGTASSRHGIQKKLKRVDQVPQKKDDSTCNSTVETRAQKARTECARKSRSEEIIDGTSEMNEGKRSQKTPSTPRRVTQGAAS
RUO
SP110
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3431
SP110 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ22835/IFI41/IFI75/IPR1/VODI
SP110
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers