missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SP1 Partial ORF (NP_612482, 89 a.a. - 198 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006667-Q01.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16084745

  • 335.00€ / 10µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

Sequence: GTGELDLTATQLSQGANGWQIISSSSGATPTSKEQSGSSTNGSNGSESSKNRTVSGGQYVVAAAPNLQNQQVLTGLPGVMPNIQYQVIPQFQTVDGQQLQFAATGAQVQQ
Specifications

Specifications

NP_612482
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GTGELDLTATQLSQGANGWQIISSSSGATPTSKEQSGSSTNGSNGSESSKNRTVSGGQYVVAAAPNLQNQQVLTGLPGVMPNIQYQVIPQFQTVDGQQLQFAATGAQVQQ
RUO
SP1
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
6667
SP1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SP1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers