missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SORCS3 Partial ORF (NP_055793, 1147 a.a. - 1222 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00022986-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16124326

  • 545.00€ / 25µg

Estimated Shipment: 06-10-2022
to see stock.
Add to basket

Description

Description

This gene encodes one family member of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins. The VPS10 domain name comes from the yeast carboxypeptidase Y sorting receptor Vps10 protein. Members of this gene family are large with many exons but the CDS lengths are usually less than 3700 nt. Very large introns typically separate the exons encoding the VPS10 domain; the remaining exons are separated by much smaller-sized introns. These genes are strongly expressed in the central nervous system. Two of the five family members (sortilin and sortilin-related receptor) are synthesized as preproproteins; it is not yet known if this encoded protein is also a preproprotein. [provided by RefSeq]

Sequence: KRKIPWINIYAQVQHDKEQEMIGSVSQSENAPKITLSDFTEPEELLDKELDTRVIGGIATIANSESTKEIPNCTSV
Specifications

Specifications

NP_055793
Liquid
22986
SORCS3 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SORCS
SORCS3
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.1kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KRKIPWINIYAQVQHDKEQEMIGSVSQSENAPKITLSDFTEPEELLDKELDTRVIGGIATIANSESTKEIPNCTSV
RUO
SORCS3
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers