Learn More
Abcam Human SNX1 knockout HeLa cell line
Product Code. 32157217 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1,000,000 Cells
2 x 1,000,000 Cells
Unit Size:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32157217 2 x 1,000,000 Cells Pack of 2
32140537 1,000,000 Cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 32157217 Supplier Abcam Supplier No. ab2738342x1000000Cells_
 Restricted Item
Estimated Shipment: 09-12-2026
SNX1 KO cell line available to order. KO validated by Next Generation Sequencing. Free of charge wild type control available. Knockout achieved by CRISPR/Cas9; X = 5 bp deletion after Lys165 of the WT protein Frameshift: 99%. To order both knockout and wild-type control cells: select 2 x 1000000...
SNX1 KO cell line available to order. KO validated by Next Generation Sequencing. Free of charge wild type control available. Knockout achieved by CRISPR/Cas9; X = 5 bp deletion after Lys165 of the WT protein Frameshift: 99%. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells