missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SNRK Partial ORF (NP_060189, 656 a.a. - 765 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00054861-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107766

  • 349.00€ / 10µg

Estimated Shipment: 07-07-2023
to see stock.
Add to basket

Description

Description

SNRK is a member of the sucrose nonfermenting (SNF)-related kinase family of serine/threonine kinases (Kertesz et al., 2002 [PubMed 12234663]).[supplied by OMIM]

Sequence: SQLHGSTKYIIDPQNGLSFSSVKVQEKSTWKMCISSTGNAGQVPAVGGIKFFSDHMADTTTELERIKSKNLKNNVLQLPLCEKTISVNIQRNPKEGLLCASSPASCCHVI
Specifications

Specifications

NP_060189
Liquid
54861
SNRK (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp779A1866/FLJ20224/HSNFRK/KIAA0096
SNRK
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SQLHGSTKYIIDPQNGLSFSSVKVQEKSTWKMCISSTGNAGQVPAVGGIKFFSDHMADTTTELERIKSKNLKNNVLQLPLCEKTISVNIQRNPKEGLLCASSPASCCHVI
RUO
SNRK
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers