Learn More
Abnova™ Human SMARCC1 Partial ORF (NP_003065, 338 a.a. - 437 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006599-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and contains a predicted leucine zipper motif typical of many transcription factors. [provided by RefSeq]
Sequence: ESRKKSGKKGQASLYGKRRSQKEEDEQEDLTKDMEDPTPVPNIEEVVLPKNVNLKKDSENTPVKGGTVADLDEQDEETVTAGGKEDEDPAKGDQSRSVDLSpecifications
NP_003065 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ESRKKSGKKGQASLYGKRRSQKEEDEQEDLTKDMEDPTPVPNIEEVVLPKNVNLKKDSENTPVKGGTVADLDEQDEETVTAGGKEDEDPAKGDQSRSVDL | |
RUO | |
SMARCC1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6599 | |
SMARCC1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BAF155/CRACC1/Rsc8/SRG3/SWI3 | |
SMARCC1 | |
Recombinant | |
wheat germ expression system |