missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SMAD5 Partial ORF (AAH09682, 105 a.a. - 182 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004090-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: KPLDICEFPFGSKQKEVCINPYHYKRVESPVLPPVLVPRHNEFNPQHSLLVQFRNLSHNEPHMPQNATFPDSFHQPNNSpecifications
AAH09682 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.21kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KPLDICEFPFGSKQKEVCINPYHYKRVESPVLPPVLVPRHNEFNPQHSLLVQFRNLSHNEPHMPQNATFPDSFHQPNN | |
RUO | |
SMAD5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4090 | |
SMAD5 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp781C1895/DKFZp781O1323/Dwfc/JV5-1/MADH5 | |
SMAD5 | |
Recombinant | |
wheat germ expression system |