missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLK Partial ORF (NP_055535, 1126 a.a. - 1235 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009748-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16101246

  • 349.00€ / 10µg

Estimated Shipment: 04-07-2023
to see stock.
Add to basket

Description

Description

Sequence: KHENQMRDLQLQCEANVRELHQLQNEKCHLLVEHETQKLKELDEEHSQELKEWREKLRPRKKTLEEEFARKLQEQEVFFKMTGESECLNPSTQSRISKFYPIPSLHSTGS
Specifications

Specifications

NP_055535
Liquid
9748
SLK (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0204/LOSK/MGC133067/STK2/bA16H23.1/se20-9
SLK
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KHENQMRDLQLQCEANVRELHQLQNEKCHLLVEHETQKLKELDEEHSQELKEWREKLRPRKKTLEEEFARKLQEQEVFFKMTGESECLNPSTQSRISKFYPIPSLHSTGS
RUO
SLK
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers