missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC9A3 Partial ORF (NP_004165.2, 670 a.a. - 779 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006550-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195415

  • 528.00€ / 25µg

Estimated Shipment: 31-10-2023
to see stock.
Add to basket

Description

Description

Sequence: GLNQNKKAAKLYKRERAQKRRNSSIPNGKLPMESPAQNFTIKEKDLELSDTEEPPNYDEEMSGGIEFLASVTKDTASDSPAGIDNPVFSPDEALDRSLLARLPPWLSPGE
Specifications

Specifications

NP_004165.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue
GLNQNKKAAKLYKRERAQKRRNSSIPNGKLPMESPAQNFTIKEKDLELSDTEEPPNYDEEMSGGIEFLASVTKDTASDSPAGIDNPVFSPDEALDRSLLARLPPWLSPGE
RUO
SLC9A3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6550
SLC9A3 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC126718/MGC126720/NHE3
SLC9A3
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers