Learn More
Abnova™ Human SLC8A3 Full-length ORF (NP_891981.1, 1 a.a. - 284 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006547-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the sodium/calcium exchanger integral membrane protein family. Three mammalian isoforms in family 8 have been identified. Na+/Ca2+ exchange proteins are involved in maintaining Ca2+ homeostasis in a wide variety of cell types. The protein is regulated by intracellular calcium ions and is found in both the plasma membrane and intracellular organellar membranes, where exchange of Na+ for Ca2+ occurs in an electrogenic manner. Alternative splicing has been observed for this gene and multiple variants have been described. [provided by RefSeq]
Sequence: MEEEEAKRIAEMGKPVLGEHPKLEVIIEESYEFKTTVDKLIKKTNLALVVGTHSWRDQFMEAITVSAAGDEDEDESGEERLPSCFDYVMHFLTVFWKVLFACVPPTEYCHGWACFAVSILIIGMLTAIIGDLASHFGCTIGLKDSVTAVVFVAFGTSVPDTFASKAAALQDVYADASIGNVTGSNAVNVFLGIGLAWSVAAIYWALQGQEFHVSAGTLAFSVTLFTIFAFVCISVLLYRRRPHLGGELGGPRGCKLATTWLFVSLWLLYILFATLEAYCYIKGFSpecifications
NP_891981.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
57.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NCX3 | |
SLC8A3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
6547 | |
SLC8A3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEEEEAKRIAEMGKPVLGEHPKLEVIIEESYEFKTTVDKLIKKTNLALVVGTHSWRDQFMEAITVSAAGDEDEDESGEERLPSCFDYVMHFLTVFWKVLFACVPPTEYCHGWACFAVSILIIGMLTAIIGDLASHFGCTIGLKDSVTAVVFVAFGTSVPDTFASKAAALQDVYADASIGNVTGSNAVNVFLGIGLAWSVAAIYWALQGQEFHVSAGTLAFSVTLFTIFAFVCISVLLYRRRPHLGGELGGPRGCKLATTWLFVSLWLLYILFATLEAYCYIKGF | |
RUO | |
SLC8A3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |