Learn More
Abnova™ Human SLC6A11 Full-length ORF (AAH36083.1, 1 a.a. - 208 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006538-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Gamma-aminobutyric acid (GABA) is a major inhibitory neurotransmitter. GABAergic neurotransmission is terminated by the uptake of GABA into the presynaptic terminal and the surrounding astroglial cells by sodium-dependent transporters, such as SLC6A11.[supplied by OMIM]
Sequence: MTAEKALPLGNGKAAEEARESEAPGGGCSSGGAAPARHPRVKRDKAVHERGHWNNKVEFVLSVAGEIIGLGNVWRFPYLCYKNGGGAFLIPYVVFFICCGIPVFFLETALGQFTSEGGITCWRKVCPLFEGIGYATQVIEAHLNVYYIIILAWAIFYLSNCFTTELPWATCGHEWNTENCVEFQKLNVSNYSHVSLQNATSPVMEFWESpecifications
AAH36083.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
49.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GAT-3/GAT3 | |
SLC6A11 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
6538 | |
SLC6A11 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTAEKALPLGNGKAAEEARESEAPGGGCSSGGAAPARHPRVKRDKAVHERGHWNNKVEFVLSVAGEIIGLGNVWRFPYLCYKNGGGAFLIPYVVFFICCGIPVFFLETALGQFTSEGGITCWRKVCPLFEGIGYATQVIEAHLNVYYIIILAWAIFYLSNCFTTELPWATCGHEWNTENCVEFQKLNVSNYSHVSLQNATSPVMEFWE | |
RUO | |
SLC6A11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |