missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC6A11 Full-length ORF (AAH36083.1, 1 a.a. - 208 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006538-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16170792

  • 508.00€ / 25µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

Gamma-aminobutyric acid (GABA) is a major inhibitory neurotransmitter. GABAergic neurotransmission is terminated by the uptake of GABA into the presynaptic terminal and the surrounding astroglial cells by sodium-dependent transporters, such as SLC6A11.[supplied by OMIM]

Sequence: MTAEKALPLGNGKAAEEARESEAPGGGCSSGGAAPARHPRVKRDKAVHERGHWNNKVEFVLSVAGEIIGLGNVWRFPYLCYKNGGGAFLIPYVVFFICCGIPVFFLETALGQFTSEGGITCWRKVCPLFEGIGYATQVIEAHLNVYYIIILAWAIFYLSNCFTTELPWATCGHEWNTENCVEFQKLNVSNYSHVSLQNATSPVMEFWE
Specifications

Specifications

AAH36083.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
49.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GAT-3/GAT3
SLC6A11
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
6538
SLC6A11 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTAEKALPLGNGKAAEEARESEAPGGGCSSGGAAPARHPRVKRDKAVHERGHWNNKVEFVLSVAGEIIGLGNVWRFPYLCYKNGGGAFLIPYVVFFICCGIPVFFLETALGQFTSEGGITCWRKVCPLFEGIGYATQVIEAHLNVYYIIILAWAIFYLSNCFTTELPWATCGHEWNTENCVEFQKLNVSNYSHVSLQNATSPVMEFWE
RUO
SLC6A11
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers