Learn More
Human SLC50A1 (RAG1AP1) knockout HeLa cell lysate, 1 Kit
Product Code. 32142499 Shop All Abcam Products
Product Code. 32142499 Supplier Abcam Supplier No. ab2591371Kit
 Restricted Item
Estimated Shipment: 10-11-2026
Human SLC50A1 (RAG1AP1) knockout HeLa cell/tissue lysate.
  • Species: Human
  • Tissue: SLC50A1 (RAG1AP1) knockout HeLa