missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLC38A4 Full-length ORF (NP_060488.2, 1 a.a. - 547 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_060488.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 55089 |
Molecular Weight (g/mol) | 87.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16101113
|
Abnova™
H00055089-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16191103
|
Abnova™
H00055089-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
SLC38A4 is found predominantly in liver and transports both cationic and neutral amino acids. The transport of cationic amino acids by SLC38A4 is Na(+) and pH independent, while the transport of neutral amino acids is Na(+) and pH dependent (Hatanaka et al., 2001 [PubMed 11342143]).[supplied by OMIM]
Sequence: MDPMELRNVNIEPDDESSSGESAPDSYIGIGNSEKAAMSSQFANEDTESQKFLTNGFLGKKKLADYADEHHPGTTSFGMSSFNLSNAIMGSGILGLSYAMANTGIILFIIMLLAVAILSLYSVHLLLKTAKEGGSLIYEKLGEKAFGWPGKIGAFVSITMQNIGAMSSYLFIIKYELPEVIRAFMGLEENTGEWYLNGNYLIIFVSVGIILPLSLLKNLGYLGYTSGFSLTCMVFFVSVVIYKKFQIPCPLPVLDHSVGNLSFNNTLPMHVVMLPNNSESSDVNFMMDYTHRNPAGLDENQAKGSLHDSGVEYEAHSDDKCEPKYFVFNSRTAYAIPILVFAFVCHPEVLPIYSELKDRSRRKMQTVSNISITGMLVMYLLAALFGYLTFYGEVEDELLHAYSKVYTLDIPLLMVRLAVLVAVTLTVPIVLFPIRTSVITLLFPKRPFSWIRHFLIAAVLIALNNVLVILVPTIKYIFGFIGASSATMLIFILPAVFYLKLVKKETFRSPQKVGALIFLVVGIFFMIGSMALIIIDWIYDPPNSKHHSpecifications
NP_060488.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
87.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MDPMELRNVNIEPDDESSSGESAPDSYIGIGNSEKAAMSSQFANEDTESQKFLTNGFLGKKKLADYADEHHPGTTSFGMSSFNLSNAIMGSGILGLSYAMANTGIILFIIMLLAVAILSLYSVHLLLKTAKEGGSLIYEKLGEKAFGWPGKIGAFVSITMQNIGAMSSYLFIIKYELPEVIRAFMGLEENTGEWYLNGNYLIIFVSVGIILPLSLLKNLGYLGYTSGFSLTCMVFFVSVVIYKKFQIPCPLPVLDHSVGNLSFNNTLPMHVVMLPNNSESSDVNFMMDYTHRNPAGLDENQAKGSLHDSGVEYEAHSDDKCEPKYFVFNSRTAYAIPILVFAFVCHPEVLPIYSELKDRSRRKMQTVSNISITGMLVMYLLAALFGYLTFYGEVEDELLHAYSKVYTLDIPLLMVRLAVLVAVTLTVPIVLFPIRTSVITLLFPKRPFSWIRHFLIAAVLIALNNVLVILVPTIKYIFGFIGASSATMLIFILPAVFYLKLVKKETFRSPQKVGALIFLVVGIFFMIGSMALIIIDWIYDPPNSKHH | |
RUO | |
SLC38A4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
55089 | |
SLC38A4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATA3/FLJ10191/MGC126876/NAT3/PAAT | |
SLC38A4 | |
Recombinant | |
wheat germ expression system |