missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLC38A2 Full-length ORF (AAH29379, 1 a.a. - 52 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00054407-P02.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MSSWKELLTLYLRPAREYQELFLQTFFFFFVWEKILWQPGRNGKKILFSTANSpecifications
AAH29379 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.46kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATA2/KIAA1382/PRO1068/SAT2/SNAT2 | |
SLC38A2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54407 | |
SLC38A2 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSSWKELLTLYLRPAREYQELFLQTFFFFFVWEKILWQPGRNGKKILFSTAN | |
RUO | |
SLC38A2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |