missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC38A2 Full-length ORF (AAH29379, 1 a.a. - 52 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00054407-P02.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100523

  • 508.00€ / 25µg
Estimated Shipment: 05-06-2024
to see stock.
Add to basket

Description

Description

Sequence: MSSWKELLTLYLRPAREYQELFLQTFFFFFVWEKILWQPGRNGKKILFSTAN
Specifications

Specifications

AAH29379
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.46kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATA2/KIAA1382/PRO1068/SAT2/SNAT2
SLC38A2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
54407
SLC38A2 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSSWKELLTLYLRPAREYQELFLQTFFFFFVWEKILWQPGRNGKKILFSTAN
RUO
SLC38A2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers