Learn More
Abnova™ Human SLC35D2 Full-length ORF (AAI00279.1, 1 a.a. - 249 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011046-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Nucleotide sugars, which are synthesized in the cytosol or the nucleus, are high-energy donor substrates for glycosyltransferases located in the lumen of the endoplasmic reticulum and Golgi apparatus. Translocation of nucleotide sugars from the cytosol into the lumen compartment is mediated by specific nucleotide sugar transporters, such as SLC35D2 (Suda et al., 2004 [PubMed 15082721]).[supplied by OMIM]
Sequence: MTAGGQAEAEGAGGEPGAARLPSRVARLLSALFYGTCSFLIVLVNKALLTTYGFPSPIFLGIGQMAATIMILYVSKLNKIIHFPDFDKKIPVKLFPLPLLYVGNHISGLSSTSKLSLPMFTVLRKFTIPLTLLLETIILGKQYSLNIILSVFAIILGAFIAAGFLLMYSTVLCSYYNSALTTAVVGAIKNVSVAYIGILIGGDYIFSLLNFVGLNICMAGGLRYSFLTLSSQLKPKPVGEENICLDLKSSpecifications
AAI00279.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
53.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HFRC1/MGC117215/MGC142139/SQV7L/UGTrel8/hfrc | |
SLC35D2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
11046 | |
SLC35D2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTAGGQAEAEGAGGEPGAARLPSRVARLLSALFYGTCSFLIVLVNKALLTTYGFPSPIFLGIGQMAATIMILYVSKLNKIIHFPDFDKKIPVKLFPLPLLYVGNHISGLSSTSKLSLPMFTVLRKFTIPLTLLLETIILGKQYSLNIILSVFAIILGAFIAAGFLLMYSTVLCSYYNSALTTAVVGAIKNVSVAYIGILIGGDYIFSLLNFVGLNICMAGGLRYSFLTLSSQLKPKPVGEENICLDLKS | |
RUO | |
SLC35D2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |