missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC2A4RG Partial ORF (NP_064446, 178 a.a. - 269 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00056731-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192263

  • 349.00€ / 10µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is a nuclear transcription factor involved in the activation of the solute carrier family 2 member 4 gene. The encoded protein interacts with another transcription factor, myocyte enhancer factor 2, to activate transcription of this gene. [provided by RefSeq]

Sequence: EAAHFLFGEPTLRKRKSPAQVMFQCLWKSCGKVLSTASAMQRHIRLVHLGRQAEPEQSDGEEDFYYTELDVGVDTLTDGLSSLTPVSPTASM
Specifications

Specifications

NP_064446
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.86kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EAAHFLFGEPTLRKRKSPAQVMFQCLWKSCGKVLSTASAMQRHIRLVHLGRQAEPEQSDGEEDFYYTELDVGVDTLTDGLSSLTPVSPTASM
RUO
SLC2A4RG
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
56731
SLC2A4RG (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GEF/HDBP1/Si-1-2/Si-1-2-19
SLC2A4RG
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers