missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC2A10 Partial ORF (NP_110404, 325 a.a. - 382 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00081031-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16160377

  • 545.00€ / 25µg

Estimated Shipment: 24-08-2022
to see stock.
Add to basket

Description

Description

SLC2A10 is a member of the facilitative glucose transporter family, which plays a significant role in maintaining glucose homeostasis.[supplied by OMIM]

Sequence: SGPSCLAVPNATGQTGLPGDSGLLQDSSLPPIPRTNEDQREPILSTAKKTKPHPRSGD
Specifications

Specifications

NP_110404
Liquid
81031
SLC2A10 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATS/GLUT10/MGC126706
SLC2A10
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.12kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SGPSCLAVPNATGQTGLPGDSGLLQDSSLPPIPRTNEDQREPILSTAKKTKPHPRSGD
RUO
SLC2A10
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers