missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC25A25 Partial ORF (NP_443133, 2 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00114789-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121417

  • 335.00€ / 10µg
Estimated Shipment: 05-06-2024
to see stock.
Add to basket

Description

Description

Sequence: LCLCLYVPVIGEAQTEFQYFESKGLPAELKSIFKLSVFIPSQEFSTYRQWKQKIVQAGDKDLDGQLDFEEFVHYLQDHEKKLRLVFKSLDKKNDGRIDAQEIMQSLRDL
Specifications

Specifications

NP_443133
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LCLCLYVPVIGEAQTEFQYFESKGLPAELKSIFKLSVFIPSQEFSTYRQWKQKIVQAGDKDLDGQLDFEEFVHYLQDHEKKLRLVFKSLDKKNDGRIDAQEIMQSLRDL
RUO
SLC25A25
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
114789
SLC25A25 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA1896/MCSC/MGC105138/MGC119514/MGC119515/MGC119516/MGC119517/PCSCL/RP11-395P17.4/SCAMC-2
SLC25A25
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers