missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC25A23 Full-length ORF (AAH01656.1, 1 a.a. - 482 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079085-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16164083

  • 545.00€ / 25µg

Estimated Shipment: 22-08-2022
to see stock.
Add to basket

Description

Description

Sequence: MRGSPGDAERRQRWGRLFEELDSNKDGRVDVHELRQGLARLGGGNPDPGAQQGISSEGDADPDGGLDLEEFSRYLQEREQRLLLMFHSLDRNQDGHIDVSEIQQSFRALGISISLEQAEKILHSMDRDGTMTIDWQEWRDHFLLHSLENVEDVLYFWKHSTLSSAGFSAWIKDSTAEQNRSKTTVLARRSGSHLKSQHFGRPKWADHEVLDIGECLTVPDEFSKQEKLTGMWWKQLVAGAVAGAVSRTGTAPLDRLKVFMQVHASKTNRLNILGGLRSMVLEGGIRSLWRGNGINVLKIAPESAIKFMAYEQIKRAILGQQETLHVQERFVAGSLAGATAQTIIYPMEVLKTRLTLRRTGQYKGLLDCARRILEREGPRAFYRGYLPNVLGIIPYAGIDLAVYETLKNWWLQQYSHDSADPGILVLLACGTISSTCGQIASYPLALVRTRMQAQGWSTVARFQITATSAFQVQAILLPQPPE
Specifications

Specifications

AAH01656.1
Liquid
79085
SLC25A23 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRGSPGDAERRQRWGRLFEELDSNKDGRVDVHELRQGLARLGGGNPDPGAQQGISSEGDADPDGGLDLEEFSRYLQEREQRLLLMFHSLDRNQDGHIDVSEIQQSFRALGISISLEQAEKILHSMDRDGTMTIDWQEWRDHFLLHSLENVEDVLYFWKHSTLSSAGFSAWIKDSTAEQNRSKTTVLARRSGSHLKSQHFGRPKWADHEVLDIGECLTVPDEFSKQEKLTGMWWKQLVAGAVAGAVSRTGTAPLDRLKVFMQVHASKTNRLNILGGLRSMVLEGGIRSLWRGNGINVLKIAPESAIKFMAYEQIKRAILGQQETLHVQERFVAGSLAGATAQTIIYPMEVLKTRLTLRRTGQYKGLLDCARRILEREGPRAFYRGYLPNVLGIIPYAGIDLAVYETLKNWWLQQYSHDSADPGILVLLACGTISSTCGQIASYPLALVRTRMQAQGWSTVARFQITATSAFQVQAILLPQPPE
RUO
SLC25A23
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
80.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
APC2/MCSC2/MGC2615/SCaMC-3
SLC25A23
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers