missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC25A22 Partial ORF (NP_078974.1, 33 a.a. - 105 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079751-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16160047

  • 359.00€ / 10µg

Estimated Shipment: 22-08-2022
to see stock.
Add to basket

Description

Description

The SLC25 gene family encodes mitochondrial carriers that transport a variety of metabolites across the inner mitochondrial membrane (Palmieri, 2004 [PubMed 14598172]). SLC25A22, also known as GC1, is 1 of the 2 mitochondrial glutamate/H+ symporters, the other being SLC25A18 (MIM 609303).[supplied by OMIM]

Sequence: KTRLQNQQNGQRVYTSMSDCLIKTVRSEGYFGMYRGAAVNLTLVTPEKAIKLAANDFFRHQLSKDGQKLTLLK
Specifications

Specifications

NP_078974.1
Liquid
79751
SLC25A22 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ13044/GC1
SLC25A22
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.77kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KTRLQNQQNGQRVYTSMSDCLIKTVRSEGYFGMYRGAAVNLTLVTPEKAIKLAANDFFRHQLSKDGQKLTLLK
RUO
SLC25A22
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers