Learn More
Abnova™ Human SLC25A15 Full-length ORF (NP_055067.1, 1 a.a. - 301 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010166-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the mitochondrial carrier family. The encoded protein transports ornithine across the inner mitochondrial membrane from the cytosol to the mitochondrial matrix. The protein is an essential component of the urea cycle, and functions in ammonium detoxification and biosynthesis of the amino acid arginine. Mutations in this gene result in hyperornithinemia-hyperammonemia-homocitrullinuria (HHH) syndrome. There is a pseudogene of this locus on the Y chromosome
Sequence: MKSNPAIQAAIDLTAGAAGGTACVLTGQPFDTMKVKMQTFPDLYRGLTDCCLKTYSQVGFRGFYKGTSPALIANIAENSVLFMCYGFCQQVVRKVAGLDKQAKLSDLQNAAAGSFASAFAALVLCPTELVKCRLQTMYEMETSGKIAKSQNTVWSVIKSILRKDGPLGFYHGLSSTLLREVPGYFFFFGGYELSRSFFASGRSKDELGPVPLMLSGGVGGICLWLAVYPVDCIKSRIQVLSMSGKQAGFIRTFINVVKNEGITALYSGLKPTMIRAFPANGALFLAYEYSRKLMMNQLEAYSpecifications
NP_055067.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
D13S327/HHH/ORC1/ORNT1 | |
SLC25A15 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10166 | |
SLC25A15 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKSNPAIQAAIDLTAGAAGGTACVLTGQPFDTMKVKMQTFPDLYRGLTDCCLKTYSQVGFRGFYKGTSPALIANIAENSVLFMCYGFCQQVVRKVAGLDKQAKLSDLQNAAAGSFASAFAALVLCPTELVKCRLQTMYEMETSGKIAKSQNTVWSVIKSILRKDGPLGFYHGLSSTLLREVPGYFFFFGGYELSRSFFASGRSKDELGPVPLMLSGGVGGICLWLAVYPVDCIKSRIQVLSMSGKQAGFIRTFINVVKNEGITALYSGLKPTMIRAFPANGALFLAYEYSRKLMMNQLEAY | |
RUO | |
SLC25A15 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |