missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC24A1 Partial ORF (NP_004718, 162 a.a. - 268 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009187-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16160116

  • 508.00€ / 25µg
Estimated Shipment: 25-06-2024
to see stock.
Add to basket

Description

Description

SLC24A1 belongs to a family of potassium-dependent sodium/calcium exchangers. Members of this family have 2 large hydrophilic loops and 2 sets of multiple transmembrane-spanning segments (Schnetkamp, 2004 [PubMed 14770312]).[supplied by OMIM]

Sequence: IVKKYTPTPRGEMKSYSPTQVREKVKYTPSPRGRRVGTYVPSTFMTMETSHAITPRTTVKDSDITATYKILETNSLKRIMEETTPTTLKGMFDSTPTFLTHEVEANV
Specifications

Specifications

NP_004718
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.51kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
IVKKYTPTPRGEMKSYSPTQVREKVKYTPSPRGRRVGTYVPSTFMTMETSHAITPRTTVKDSDITATYKILETNSLKRIMEETTPTTLKGMFDSTPTFLTHEVEANV
RUO
SLC24A1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9187
SLC24A1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HsT17412/KIAA0702/NCKX/NCKX1/RODX
SLC24A1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers