Learn More
Abnova™ Human SLC22A3 Partial ORF (NP_068812, 90 a.a. - 155 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006581-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. [provided by RefSeq]
Sequence: CQRYLLEAANDSASATSALSCADPLAAFPNRSAPLVPCRGGWRYAQAHSTIVSEFDLVCVNAWMLDSpecifications
NP_068812 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
CQRYLLEAANDSASATSALSCADPLAAFPNRSAPLVPCRGGWRYAQAHSTIVSEFDLVCVNAWMLD | |
RUO | |
SLC22A3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6581 | |
SLC22A3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EMT/EMTH/OCT3 | |
SLC22A3 | |
Recombinant | |
wheat germ expression system |