missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SIK2 Partial ORF (NP_056006, 467 a.a. - 576 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023235-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: AHAFEAFQSTRSGQRRHTLSEVTNQLVVMPGAGKIFSMNDSPSLDSVDSEYDMGSVQRDLNFLEDNPSLKDIMLANQPSPRMTSPFISLRPTNPAMQALSSQKREVHNRSSpecifications
NP_056006 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AHAFEAFQSTRSGQRRHTLSEVTNQLVVMPGAGKIFSMNDSPSLDSVDSEYDMGSVQRDLNFLEDNPSLKDIMLANQPSPRMTSPFISLRPTNPAMQALSSQKREVHNRS | |
RUO | |
SIK2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23235 | |
SIK2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434K1115/KIAA0781/LOH11CR1I/QIK/SNF1LK2 | |
SIK2 | |
Recombinant | |
wheat germ expression system |