Learn More
R&D Systems™ Human Siglec-8 Fc Chimera, Carrier-Free Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 9045-SL-050
Description
The Recombinant Human Siglec-8 Fc Chimera Protein is derived from NS0. The Recombinant Human Siglec-8 Fc Chimera Protein has been validated for the following applications: Bioactivity.Specifications
| AAH53319 | |
| Observed M.W.: 68-88kDa, reducing conditions, Theoretical M.W.: 64kDa | |
| Mouse myeloma cell line,NS0-derived human Siglec-8 protein, (N-terminus) Human Siglec-8 (Met17-Ala363) Accession &Num; AAH53319 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) | |
| <0.10 EU per 1μg of the protein by the LAL method. | |
| Siglec-8 | |
| Measured by its binding ability in a functional ELISA. When Biotinylated Sialyl Lewis X was immobilized at 0.05μg/mL (100μL/well), it binds Recombinant Human Siglec-8 Fc Chimera. The concentration of Human Siglec-8 Fc Chimera that produces 50% of the optimal binding response is approximately 0.2-1.2μg/mL. | |
| Yes |
| 27181 | |
| 50μg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
| CD329 antigen, CDw329, MGC59785, SAF2, SAF2SAF-2, sialic acid binding Ig-like lectin 8, Sialoadhesin family member 2, Siglec8, Siglec-8, SIGLEC8L, SIGLEC8Lsialic acid-binding Ig-like lectin 8 | |
| SIGLEC8 | |
| Unconjugated |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.