missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Siglec-2/CD22 DuoSet ELISA

Product Code. 30001725
Click to view available options
Sufficient For:
Five 96-well plates
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 30001725

Brand: R&D Systems™ DY196805

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For determination of natural and recombinant human Sialic Acid Binding Ig-like Lec protein.

The Human Siglec-2/CD22 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Siglec-2/CD22.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 93.80 to 6,000pg/mL
Immunoassay Kit Format 96-well Plate
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Sufficient For Five 96-well plates
Target Species Human
Gene ID (Entrez) 933 (Human); 12483 (Mouse)
Kit Contents 96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Target Sialic Acid Binding Ig-like Lectin 2

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.