missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SHFM1 Full-length ORF (NP_006295.1, 1 a.a. - 70 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007979-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16124568

  • 508.00€ / 25µg
Estimated Shipment: 07-05-2024
to see stock.
Add to basket

Description

Description

The product of this gene has been localized within the split hand/split foot malformation locus SHFM1 at chromosome 7. It has been proposed to be a candidate gene for the autosomal dominant form of the heterogeneous limb developmental disorder split hand/split foot malformation type 1. In addition, it has been shown to directly interact with BRCA2. It also may play a role in the completion of the cell cycle. [provided by RefSeq]

Sequence: MSEKKQPVDLGLLEEDDEFEEFPAEDWAGLDEDEDAHVWEDNWDDDNVEDDFSNQLRAELEKHGYKMETS
Specifications

Specifications

NP_006295.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DSS1/ECD/SEM1/SHFD1/SHSF1/Shfdg1
SHFM1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
7979
SHFM1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSEKKQPVDLGLLEEDDEFEEFPAEDWAGLDEDEDAHVWEDNWDDDNVEDDFSNQLRAELEKHGYKMETS
RUO
SHFM1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers