Learn More
R&D Systems™ Human SHBG Quantikine ELISA Kit
Product Code. 18718838
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. 18718838
Brand: R&D Systems™ DSHBG0B
Human SHBG Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Human SHBG Quantikine ELISA Kit quantifies human SHBG in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).
TRUSTED_SUSTAINABILITY
Specifications
Assay Range 0.06 - 4 nmol/L
Assay Sensitivity 0.012 nmol/L
Detectable Analytes SHBG
Assay Solid Phase ELISA
Conjugate HRP
Cross Reactivity <0.5% cross-reactivity observed with available related molecules.Cross-species reactivity not tested
Immunoassay Kit Format 96-well Strip Plate
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity Natural and Recombinant Human SHBG
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 6462
Host Species Human
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym ABP, ABPTestosterone-estrogen-binding globulin, androgen binding protein, MGC126834, MGC138391, SBP, sex hormone-binding globulin, Sex steroid-binding protein, TeBG, Testis-specific androgen-binding protein, testosterone-binding beta-globulin, Testosterone-estradiol-binding globulin
Total Assay Time 4.5 hr.