missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SH3GL1 Full-length ORF (NP_003016.1, 1 a.a. - 368 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006455-P02.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195205

  • 349.00€ / 10µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

Sequence: MSVAGLKKQFYKASQLVSEKVGGAEGTKLDDDFKEMEKKVDVTSKAVTEVLARTIEYLQPNPASRAKLTMLNTVSKIRGQVKNPGYPQSEGLLGECMIRHGKELGGESNFGDALLDAGESMKRLAEVKDSLDIEVKQNFIDPLQNLCEKDLKEIQHHLKKLEGRRLDFDYKKKRQGKIPDEELRQALEKFEESKEVAETSMHNLLETDIEQVSQLSALVDAQLDYHRQAVQILDELAEKLKRRMREASSRPKREYKPKPREPFDLGEPEQSNGGFPCTTAPKIAASSSFRSSDKPIRTPSRSMPPLDQPSCKALYDFEPENDGELGFHEGDVITLTNQIDENWYEGMLDGQSGFFPLSYVEVLVPLPQ
Specifications

Specifications

NP_003016.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
67.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CNSA1/EEN/MGC111371/SH3D2B/SH3P8
SH3GL1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
6455
SH3GL1 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSVAGLKKQFYKASQLVSEKVGGAEGTKLDDDFKEMEKKVDVTSKAVTEVLARTIEYLQPNPASRAKLTMLNTVSKIRGQVKNPGYPQSEGLLGECMIRHGKELGGESNFGDALLDAGESMKRLAEVKDSLDIEVKQNFIDPLQNLCEKDLKEIQHHLKKLEGRRLDFDYKKKRQGKIPDEELRQALEKFEESKEVAETSMHNLLETDIEQVSQLSALVDAQLDYHRQAVQILDELAEKLKRRMREASSRPKREYKPKPREPFDLGEPEQSNGGFPCTTAPKIAASSSFRSSDKPIRTPSRSMPPLDQPSCKALYDFEPENDGELGFHEGDVITLTNQIDENWYEGMLDGQSGFFPLSYVEVLVPLPQ
RUO
SH3GL1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers