Learn More
Abnova™ Human SH2B Full-length ORF (AAH10704.1, 1 a.a. - 426 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00025970-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the SH2-domain containing mediators family. The encoded protein mediates activation of various kinases and may function in cytokine and growth factor receptor signaling and cellular transformation. Alternatively spliced transcript variants have been described. [provided by RefSeq]
Sequence: MVQREELLSFMGAEEAAPDPAGVGRGGGVAGPPSGGGGQPQWQKCRLLLRSEGEGGGGSRLEFFVPPKASRPRLSIPCSSITDVRTTTALEMPDRENTFVVKVEGPSEYIMETVDAQHVKAWVSDIQECLSPGPCPATSPRPMTLPLAPGTSFLTRENTDSLELSCLNHSESLPSQDLLLGPSESNDRLSQGAYGGLSDRPSASISPSSASIAASHFDSMELLPPELPPRIPIEEGPPAGTVHPLSAPYPPLDTPETATGSFLFQGEPEGGEGDQPLSGYPWFHGMLSRLKAAQLALTGGTGSHGVFLVRQSETRRGEYVLTFNFQGKAKHLRLSLNEEGQCRVQHLWFQSIFDMLEHFRVHPIPLESGGSSDVVLVSYVPSSQRQQGREQAGSHAGVCEGDGCHPDASCTLMPFGASDCVTDHLPSpecifications
AAH10704.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
71.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp547G1110/FLJ30542/KIAA1299/SH2-B/SH2B | |
SH2B1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
25970 | |
SH2B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVQREELLSFMGAEEAAPDPAGVGRGGGVAGPPSGGGGQPQWQKCRLLLRSEGEGGGGSRLEFFVPPKASRPRLSIPCSSITDVRTTTALEMPDRENTFVVKVEGPSEYIMETVDAQHVKAWVSDIQECLSPGPCPATSPRPMTLPLAPGTSFLTRENTDSLELSCLNHSESLPSQDLLLGPSESNDRLSQGAYGGLSDRPSASISPSSASIAASHFDSMELLPPELPPRIPIEEGPPAGTVHPLSAPYPPLDTPETATGSFLFQGEPEGGEGDQPLSGYPWFHGMLSRLKAAQLALTGGTGSHGVFLVRQSETRRGEYVLTFNFQGKAKHLRLSLNEEGQCRVQHLWFQSIFDMLEHFRVHPIPLESGGSSDVVLVSYVPSSQRQQGREQAGSHAGVCEGDGCHPDASCTLMPFGASDCVTDHLP | |
RUO | |
SH2B1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |