missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SF3A1 Partial ORF (NP_005868, 140 a.a. - 227 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010291-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192306

  • 349.00€ / 10µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

This gene encodes subunit 1 of the splicing factor 3a protein complex. The splicing factor 3a heterotrimer includes subunits 1, 2 and 3 and is necessary for the in vitro conversion of 15S U2 snRNP into an active 17S particle that performs pre-mRNA splicing. Subunit 1 belongs to the SURP protein family; named for the SURP (also called SWAP or Suppressor-of-White-APricot) motifs that are thought to mediate RNA binding. Subunit 1 has tandemly repeated SURP motifs in its amino-terminal half while its carboxy-terminal half contains a proline-rich region and a ubiquitin-like domain. Binding studies with truncated subunit 1 derivatives demonstrated that the two SURP motifs are necessary for binding to subunit 3 while contacts with subunit 2 may occur through sequences carboxy-terminal to the SURP motifs. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: TIVPKEPPPEFEFIADPPSISAFDLDVVKLTAQFVARNGRQFLTQLMQKEQRNYQFDFLRPQHSLFNYFTKLVEQYTKILIPPKGLFS
Specifications

Specifications

NP_005868
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.42kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TIVPKEPPPEFEFIADPPSISAFDLDVVKLTAQFVARNGRQFLTQLMQKEQRNYQFDFLRPQHSLFNYFTKLVEQYTKILIPPKGLFS
RUO
SF3A1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10291
SF3A1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PRP21/PRPF21/SAP114/SF3A120
SF3A1
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers