Learn More
Abcam Human SERPINE1 knockout U-87 MG cell line
Description
SERPINE1 KO cell line available to order. KO validated by Next Generation Sequencing, Western blot. Free of charge wild type control available. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Quantity | 1,000,000 Cells |
| Regulatory Status | RUO |