missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SERPINB9 Partial ORF (NP_004146, 279 a.a. - 375 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005272-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192765

  • 528.00€ / 25µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

PI9 belongs to the large superfamily of serine proteinase inhibitors (serpins), which bind to and inactivate serine proteinases. These interactions are involved in many cellular processes, including coagulation, fibrinolysis, complement fixation, matrix remodeling, and apoptosis (Sprecher et al., 1995 [PubMed 8530382]).[supplied by OMIM]

Sequence: DMESVLRHLGIVDAFQQGKADLSAMSAERDLCLSKFVHKSFVEVNEEGTEAAAASSCFVVAECCMESGPRFCADHPFLFFIRHNRANSILFCGRFSS
Specifications

Specifications

NP_004146
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.41kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DMESVLRHLGIVDAFQQGKADLSAMSAERDLCLSKFVHKSFVEVNEEGTEAAAASSCFVVAECCMESGPRFCADHPFLFFIRHNRANSILFCGRFSS
RUO
SERPINB9
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5272
SERPINB9 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAP-3/CAP3/PI9
SERPINB9
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers