missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SERF1A Full-length ORF (NP_068802.1, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008293-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16102352

  • 335.00€ / 10µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

This gene is part of a 500 kb inverted duplication on chromosome 5q13. This duplicated region contains at least four genes and repetitive elements which make it prone to rearrangements and deletions. The repetitiveness and complexity of the sequence have also caused difficulty in determining the organization of this genomic region. The duplication region includes both a telomeric and a centromeric copy of this gene. Deletions of this gene, the telomeric copy, often accompany deletions of the neighboring SMN1 gene in spinal muscular atrophy (SMA) patients, and so it is thought that this gene may be a modifier of the SMA phenotype. The function of this protein is not known; however, it bears low-level homology with the RNA-binding domain of matrin-cyclophilin, a protein which colocalizes with small nuclear ribonucleoproteins (snRNPs) and the SMN1 gene product. Alternatively spliced transcripts have been documented but it is unclear whether alternative splicing occurs for both the centromeric and telomeric copies of the gene. [provided by RefSeq]

Sequence: MARGNQRELARQKNMKKTQEISKGKRKEDSLTASQRKQSSGGQKSESKMSAGPHLPLKAPRENPCFPLPAAGGSRYYLAYGSITPISAFVFVVFFSVFFPSFYEDFCCWI
Specifications

Specifications

NP_068802.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
38.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
4F5/FAM2A/H4F5/SERF1/SMAM1
SERF1A
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
8293
SERF1A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MARGNQRELARQKNMKKTQEISKGKRKEDSLTASQRKQSSGGQKSESKMSAGPHLPLKAPRENPCFPLPAAGGSRYYLAYGSITPISAFVFVVFFSVFFPSFYEDFCCWI
RUO
SERF1A
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers