Learn More
Abnova™ Human SEPT4 Full-length ORF (NP_004565.1, 1 a.a. - 478 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005414-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the septin gene family of nucleotide binding proteins, originally described in yeast as cell division cycle regulatory proteins. Septins are highly conserved in yeast, Drosophila, and mouse and appear to regulate cytoskeletal organization. The protein encoded by this gene is thought to be part of a complex involved in cytokinesis. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. [provided by RefSeq]
Sequence: MDRSLGWQGNSVPEDRTEAGIKRFLEDTTDDGELSKFVKDFSGNASCHPPEAKTWASRPQVPEPRPQAPDLYDDDLEFRPPSRPQSSDNQQYFCAPAPLSPSARPRSPWGKLDPYDSSEDDKEYVGFATLPNQVHRKSVKKGFDFTLMVAGESGLGKSTLVNSLFLTDLYRDRKLLGAEERIMQTVEITKHAVDIEEKGVRLRLTIVDTPGFGDAVNNTECWKPVAEYIDQQFEQYFRDESGLNRKNIQDNRVHCCLYFISPFGHGLRPLDVEFMKALHQRVNIVPILAKADTLTPPEVDHKKRKIREEIEHFGIKIYQFPDCDSDEDEDFKLQDQALKESIPFAVIGSNTVVEARGRRVRGRLYPWGIVEVENPGHCDFVKLRTMLVRTHMQDLKDVTRETHYENYRAQCIQSMTRLVVKERNRNKLTRESGTDFPIPAVPPGTDPETEKLIREKDEELRRMQEMLHKIQKQMKENYSpecifications
NP_004565.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
81.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARTS/BRADEION/CE5B3/H5/MART/PNUTL2/SEP4/hCDCREL-2/hucep-7 | |
SEPT4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5414 | |
SEPT4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDRSLGWQGNSVPEDRTEAGIKRFLEDTTDDGELSKFVKDFSGNASCHPPEAKTWASRPQVPEPRPQAPDLYDDDLEFRPPSRPQSSDNQQYFCAPAPLSPSARPRSPWGKLDPYDSSEDDKEYVGFATLPNQVHRKSVKKGFDFTLMVAGESGLGKSTLVNSLFLTDLYRDRKLLGAEERIMQTVEITKHAVDIEEKGVRLRLTIVDTPGFGDAVNNTECWKPVAEYIDQQFEQYFRDESGLNRKNIQDNRVHCCLYFISPFGHGLRPLDVEFMKALHQRVNIVPILAKADTLTPPEVDHKKRKIREEIEHFGIKIYQFPDCDSDEDEDFKLQDQALKESIPFAVIGSNTVVEARGRRVRGRLYPWGIVEVENPGHCDFVKLRTMLVRTHMQDLKDVTRETHYENYRAQCIQSMTRLVVKERNRNKLTRESGTDFPIPAVPPGTDPETEKLIREKDEELRRMQEMLHKIQKQMKENY | |
RUO | |
SEPT4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |