Learn More
Abnova™ Human SEMA4G Full-length ORF (AAH20960, 1 a.a. - 127 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00057715-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Semaphorins are a large family of conserved secreted and membrane associated proteins which possess a semaphorin (Sema) domain and a PSI domain (found in plexins, semaphorins and integrins) in the N-terminal extracellular portion. Based on sequence and structural similarities, semaphorins are put into eight classes: invertebrates contain classes 1 and 2, viruses have class V, and vertebrates contain classes 3-7. Semaphorins serve as axon guidance ligands via multimeric receptor complexes, some (if not all) containing plexin proteins. This gene encodes a class 4 semaphorin. This gene and the gene for mitochondrial ribosomal protein L43 overlap at map location 10q24.31 and are transcribed in opposite directions. [provided by RefSeq]
Sequence: MGSMSPPSAWPCVLDGPETRQDLCQPPKPCVHSHAHMEECLSAGLQCPHPHLLLVHSCFIPASGLGVPSQLPHPIWSSSPASCGDLFVKSLGTGQPGEVRLHHSPPLPSCVALVNQPPHSPWSFSRVSpecifications
AAH20960 | |
Liquid | |
57715 | |
SEMA4G (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGSMSPPSAWPCVLDGPETRQDLCQPPKPCVHSHAHMEECLSAGLQCPHPHLLLVHSCFIPASGLGVPSQLPHPIWSSSPASCGDLFVKSLGTGQPGEVRLHHSPPLPSCVALVNQPPHSPWSFSRV | |
RUO | |
SEMA4G | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
39.71kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20590/KIAA1619/MGC102867 | |
SEMA4G | |
Yes | |
wheat germ expression system |