missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SEMA4G Full-length ORF (AAH20960.1, 1 a.a. - 127 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00057715-P02.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122943

  • 359.00€ / 10µg

Estimated Shipment: 27-09-2022
to see stock.
Add to basket

Description

Description

Semaphorins are a large family of conserved secreted and membrane associated proteins which possess a semaphorin (Sema) domain and a PSI domain (found in plexins, semaphorins and integrins) in the N-terminal extracellular portion. Based on sequence and structural similarities, semaphorins are put into eight classes: invertebrates contain classes 1 and 2, viruses have class V, and vertebrates contain classes 3-7. Semaphorins serve as axon guidance ligands via multimeric receptor complexes, some (if not all) containing plexin proteins. This gene encodes a class 4 semaphorin. This gene and the gene for mitochondrial ribosomal protein L43 overlap at map location 10q24.31 and are transcribed in opposite directions. [provided by RefSeq]

Sequence: MGSMSPPSAWPCVLDGPETRQDLCQPPKPCVHSHAHMEECLSAGLQCPHPHLLLVHSCFIPASGLGVPSQLPHPIWSSSPAPCGDLFVKSLGTGQPGEVRLHHSPPLPSCVALVNQPPHSPWSFSRV
Specifications

Specifications

AAH20960.1
Liquid
57715
SEMA4G (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGSMSPPSAWPCVLDGPETRQDLCQPPKPCVHSHAHMEECLSAGLQCPHPHLLLVHSCFIPASGLGVPSQLPHPIWSSSPAPCGDLFVKSLGTGQPGEVRLHHSPPLPSCVALVNQPPHSPWSFSRV
RUO
SEMA4G
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
39.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ20590/KIAA1619/MGC102867
SEMA4G
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers