missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SEC23A Full-length ORF (AAH36649.1, 1 a.a. - 765 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010484-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16165182

  • 508.00€ / 25µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is a member of the SEC23 subfamily of the SEC23/SEC24 family. It is part of a protein complex and found in the ribosome-free transitional face of the endoplasmic reticulum (ER) and associated vesicles. This protein has similarity to yeast Sec23p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. The encoded protein is suggested to play a role in the ER-Golgi protein trafficking. [provided by RefSeq]

Sequence: MTTYLEFIQQNEERDGVRFSWNVWPSSRLEATRMVVPVAALFTPLKERPDLPPIQYEPVLCSRTTCRAVLNPLCQVDYRAKLWACNFCYQRNQFPPSYAGISELNQPAELLPQFSSIEYVVLRGPQMPLIFLYVVDTCMEDEDLQALKESMQMSLSLLPPTALVGLITFGRMVQVHELGCEGISKSYVFRGTKDLSAKQLQEMLGLSKVPVTQATRGPQVQQPPPSNRFLQPVQKIDMNLTDLLGELQRDPWPVPQGKRPLRSSGVALSIAVGLLECTFPNTGARIMMFIGGPATQGPGMVVGDELKTPIRSWHDIDKDNAKYVKKGTKHFEALANRAATTGHVIDIYACALDQTGLLEMKCCPNLTGGYMVMGDSFNTSLFKQTFQRVFTKDMHGQFKMGFGGTLEIKTSREIKISGAIGPCVSLNSKGPCVSENEIGTGGTCQWKICGLSPTTTLAIYFEVVNQHNAPIPQGGRGAIQFVTQYQHSSGQRRIRVTTIARNWADAQTQIQNIAASFDQEAAAILMARLAIYRAETEEGPDVLRWLDRQLIRLCQKFGEYHKDDPSSFRFSETFSLYPQFMFHLRRSSFLQVFNNSPDESSYYRHHFMRQDLTQSLIMIQPIMYAYSFSGPPEPVLLDSSSILADRILLMDTFFQILIYHGETIAQWRKSGYQDMPEYENFRHLLQAPVDDAQEILHSRFPMPRYIDTEHGGSQARFLLSKVNPSQTHNNMYAWGQESGAPILTDDVSLQVFMDHLKKLAVSSAA
Specifications

Specifications

AAH36649.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
112.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CLSD/MGC26267
SEC23A
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10484
SEC23A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTTYLEFIQQNEERDGVRFSWNVWPSSRLEATRMVVPVAALFTPLKERPDLPPIQYEPVLCSRTTCRAVLNPLCQVDYRAKLWACNFCYQRNQFPPSYAGISELNQPAELLPQFSSIEYVVLRGPQMPLIFLYVVDTCMEDEDLQALKESMQMSLSLLPPTALVGLITFGRMVQVHELGCEGISKSYVFRGTKDLSAKQLQEMLGLSKVPVTQATRGPQVQQPPPSNRFLQPVQKIDMNLTDLLGELQRDPWPVPQGKRPLRSSGVALSIAVGLLECTFPNTGARIMMFIGGPATQGPGMVVGDELKTPIRSWHDIDKDNAKYVKKGTKHFEALANRAATTGHVIDIYACALDQTGLLEMKCCPNLTGGYMVMGDSFNTSLFKQTFQRVFTKDMHGQFKMGFGGTLEIKTSREIKISGAIGPCVSLNSKGPCVSENEIGTGGTCQWKICGLSPTTTLAIYFEVVNQHNAPIPQGGRGAIQFVTQYQHSSGQRRIRVTTIARNWADAQTQIQNIAASFDQEAAAILMARLAIYRAETEEGPDVLRWLDRQLIRLCQKFGEYHKDDPSSFRFSETFSLYPQFMFHLRRSSFLQVFNNSPDESSYYRHHFMRQDLTQSLIMIQPIMYAYSFSGPPEPVLLDSSSILADRILLMDTFFQILIYHGETIAQWRKSGYQDMPEYENFRHLLQAPVDDAQEILHSRFPMPRYIDTEHGGSQARFLLSKVNPSQTHNNMYAWGQESGAPILTDDVSLQVFMDHLKKLAVSSAA
RUO
SEC23A
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers