Learn More
R&D Systems™ Human SCD5 (NP_001032671.2) VersaClone cDNA
Product Code. 18720389
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18720389 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18720389 Supplier R&D Systems™ Supplier No. RDC3005
  • ONLINE EXCLUSIVE PRICE
    341.00€
    322.35€ / 10µg
    Save 18.65€
    5% Off
 Restricted Item
Estimated Shipment: 22-10-2026
Contains gene inserts that are fully sequenced and produced with high quality
TRUSTED_SUSTAINABILITY
Specifications
Gene SCD5
Accession Number NP_001032671.2
Gene ID (Entrez) 79966
Gene Alias ACOD4, ACOD4Acyl-CoA-desaturase 4, FADS4, FLJ21032, HSCD5, HSCD5Stearoyl-CoA 9-desaturase, SCD2, SCD4, SCD4EC 1.14.19.1, stearoyl-CoA desaturase 4, stearoyl-CoA desaturase 5
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Product Line R&D Systems
Quantity 10 μg