missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SC5DL Full-length ORF (NP_001020127.1, 1 a.a. - 299 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006309-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122548

  • 508.00€ / 25µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes an enzyme of cholesterol biosynthesis. The encoded protein catalyzes the conversion of lathosterol into 7-dehydrocholesterol. Mutations in this gene have been associated with lathosterolosis. Alternatively spliced transcript variants encoding the same protein have been described. [provided by RefSeq]

Sequence: MDLVLRVADYYFFTPYVYPATWPEDDIFRQAISLLIVTNVGAYILYFFCATLSYYFVFDHALMKHPQFLKNQVRREIKFTVQALPWISILTVALFLLEIRGYSKLHDDLGEFPYGLFELVVSIISFLFFTDMFIYWIHRGLHHRLVYKRLHKPHHIWKIPTPFASHAFHPIDGFLQSLPYHIYPFIFPLHKVVYLSLYILVNIWTISIHDGDFRVPQILQPFINGSAHHTDHHMFFDYNYGQYFTLWDRIGGSFKNPSSFEGKGPLSYVKEMTEGKRSSHSGNGCKNEKLFNGEFTKTE
Specifications

Specifications

NP_001020127.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
61.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ERG3/S5DES/SC5D
SC5DL
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
6309
SC5DL (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDLVLRVADYYFFTPYVYPATWPEDDIFRQAISLLIVTNVGAYILYFFCATLSYYFVFDHALMKHPQFLKNQVRREIKFTVQALPWISILTVALFLLEIRGYSKLHDDLGEFPYGLFELVVSIISFLFFTDMFIYWIHRGLHHRLVYKRLHKPHHIWKIPTPFASHAFHPIDGFLQSLPYHIYPFIFPLHKVVYLSLYILVNIWTISIHDGDFRVPQILQPFINGSAHHTDHHMFFDYNYGQYFTLWDRIGGSFKNPSSFEGKGPLSYVKEMTEGKRSSHSGNGCKNEKLFNGEFTKTE
RUO
SC5DL
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers