missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SAE1 Full-length ORF (AAH18271, 1 a.a. - 346 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010055-P02.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194422

  • 349.00€ / 10µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

Sequence: MVEKEEAGGGISEEEAAQYDRQIRLWGLEAQKRLRASRVLLVGLKGLGAEIAKNLILAGVKGLTMLDHEQVTPEDPGAQFLIRTGSVGRNRAEASLERAQNLNPMVDVKVDTEDIEKKPESFFTQFDAVCLTCCSRDVIVKVDQICHKNSIKFFTGDVFGYHGYTFANLGEHEFVEEKTKVAKVSQGVEDGPDTKRAKLDSSETTMVKKKVVFCPVKEALEVDWSSEKAKAALKRTTSDYFLLQVLLKFRTDKGRDPSSDTYEEDSELLLQIRNDVLDSLGISPDLLPEDFVRYCFSEMAPVCAVVGGILAQEIVKALSQRDPPHNNFFFFDGMKGNGIVECLGPK
Specifications

Specifications

AAH18271
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
63.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AOS1/FLJ3091/HSPC140/SUA1
SAE1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10055
SAE1 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVEKEEAGGGISEEEAAQYDRQIRLWGLEAQKRLRASRVLLVGLKGLGAEIAKNLILAGVKGLTMLDHEQVTPEDPGAQFLIRTGSVGRNRAEASLERAQNLNPMVDVKVDTEDIEKKPESFFTQFDAVCLTCCSRDVIVKVDQICHKNSIKFFTGDVFGYHGYTFANLGEHEFVEEKTKVAKVSQGVEDGPDTKRAKLDSSETTMVKKKVVFCPVKEALEVDWSSEKAKAALKRTTSDYFLLQVLLKFRTDKGRDPSSDTYEEDSELLLQIRNDVLDSLGISPDLLPEDFVRYCFSEMAPVCAVVGGILAQEIVKALSQRDPPHNNFFFFDGMKGNGIVECLGPK
RUO
SAE1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers