missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RSC1A1 Partial ORF (NP_006502, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006248-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194795

  • 349.00€ / 10µg

Estimated Shipment: 31-10-2023
to see stock.
Add to basket

Description

Description

Sequence: MSSLPTSDGFNHPARSSGQSPDVGNPMSLARSVSASVCPIKPSDSDRIEPKAVKALKASAEFQLNSEKKEHLSLQDLSDHASSADHAPTDQSPAMPMQNS
Specifications

Specifications

NP_006502
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MSSLPTSDGFNHPARSSGQSPDVGNPMSLARSVSASVCPIKPSDSDRIEPKAVKALKASAEFQLNSEKKEHLSLQDLSDHASSADHAPTDQSPAMPMQNS
RUO
RSC1A1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6248
RSC1A1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RS1
RSC1A1
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers