missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RRN3 Full-length ORF (ENSP00000219758, 1 a.a. - 106 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00054700-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100703

  • 335.00€ / 10µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

Sequence: MRALENDFFNSPPRKTVRFGGTVTEVLLKYKKGETNDFELLKNQLLDPDIKDDQIINWLLEFRSSVMYLTKDFEQLISIILRLPWLNRSQTVVEEYLAFLGNLVSA
Specifications

Specifications

ENSP00000219758
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
38.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp566E104/MGC104238/TIFIA
RRN3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
54700
RRN3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRALENDFFNSPPRKTVRFGGTVTEVLLKYKKGETNDFELLKNQLLDPDIKDDQIINWLLEFRSSVMYLTKDFEQLISIILRLPWLNRSQTVVEEYLAFLGNLVSA
RUO
RRN3
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers