Learn More
Abnova™ Human RRAGD Full-length ORF (NP_067067.1, 1 a.a. - 400 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00058528-P01.25ug
Additional Details : Weight : 0.00010kg
Description
RRAGD is a monomeric guanine nucleotide-binding protein, or G protein. By binding GTP or GDP, small G proteins act as molecular switches in numerous cell processes and signaling pathways.[supplied by OMIM]
Sequence: MSQVLGKPQPQDEDDAEEEEEEDELVGLADYGDGPDSSDADPDSGTEEGVLDFSDPFSTEVKPRILLMGLRRSGKSSIQKVVFHKMSPNETLFLESTNKICREDVSNSSFVNFQIWDFPGQIDFFDPTFDYEMIFRGTGALIFVIDSQDDYMEALARLHLTVTRAYKVNTDINFEVFIHKVDGLSDDHKIETQRDIHQRANDDLADAGLEKIHLSFYLTSIYDHSIFEAFSKVVQKLIPQLPTLENLLNIFISNSGIEKAFLFDVVSKIYIATDSTPVDMQTYELCCDMIDVVIDISCIYGLKEDGAGTPYDKESTAIIKLNNTTVLYLKEVTKFLALVCFVREESFERKGLIDYNFHCFRKAIHEVFEVRMKVVKSRKVQNRLQKKKRATPNGTPRVLLSpecifications
NP_067067.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
72kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp761H171/FLJ44176/RAGD/bA11D8.2.1 | |
RRAGD | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
58528 | |
RRAGD (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSQVLGKPQPQDEDDAEEEEEEDELVGLADYGDGPDSSDADPDSGTEEGVLDFSDPFSTEVKPRILLMGLRRSGKSSIQKVVFHKMSPNETLFLESTNKICREDVSNSSFVNFQIWDFPGQIDFFDPTFDYEMIFRGTGALIFVIDSQDDYMEALARLHLTVTRAYKVNTDINFEVFIHKVDGLSDDHKIETQRDIHQRANDDLADAGLEKIHLSFYLTSIYDHSIFEAFSKVVQKLIPQLPTLENLLNIFISNSGIEKAFLFDVVSKIYIATDSTPVDMQTYELCCDMIDVVIDISCIYGLKEDGAGTPYDKESTAIIKLNNTTVLYLKEVTKFLALVCFVREESFERKGLIDYNFHCFRKAIHEVFEVRMKVVKSRKVQNRLQKKKRATPNGTPRVLL | |
RUO | |
RRAGD | |
Wheat Germ (in vitro) | |
GST | |
Liquid |