Learn More
Abnova™ Human RPP21 Full-length ORF (AAH11730.1, 1 a.a. - 154 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00079897-P01.10ug
Additional Details : Weight : 0.00010kg
Description
RPP21 is a protein subunit of nuclear ribonuclease P, which processes the 5-prime leader sequence of precursor tRNAs (Jarrous et al., 2001 [PubMed 11497433]).[supplied by OMIM]
Sequence: MAGPVKDREAFQRLNFLYQAAHCVLAQDPENQALARFYCYTERTIAKRLVLRRDPSVKRTLCRGCSSLLVPGLTCTHRQRRCRGQRWTVQTCLTCQRSQRFLNDPGHLLWGDRPEAQLGSQADSKPLQPLPNTAHSISDRLPEEKMQTQGSSNQSpecifications
AAH11730.1 | |
Liquid | |
79897 | |
RPP21 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAGPVKDREAFQRLNFLYQAAHCVLAQDPENQALARFYCYTERTIAKRLVLRRDPSVKRTLCRGCSSLLVPGLTCTHRQRRCRGQRWTVQTCLTCQRSQRFLNDPGHLLWGDRPEAQLGSQADSKPLQPLPNTAHSISDRLPEEKMQTQGSSNQ | |
RUO | |
RPP21 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
44kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C6orf135/FLJ22638 | |
RPP21 | |
Yes | |
wheat germ expression system |