Learn More
Abnova™ Human RPL18A Full-length ORF (NP_000971.1, 1 a.a. - 176 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006142-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L18AE family of ribosomal proteins. It is located in the cytoplasm. This gene is co-transcribed with the U68 snoRNA, which is located in its third intron. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq]
Sequence: MKASGTLREYKVVGRCLPTPKCHTPPLYRMRIFAPNHVVAKSRFWYFVSQLKKMKKSSGEIVYCGQVFEKSPLRVKNFGIWLRYDSRSGTHNMYREYRDLTTAGAVTQCYRDMGARHRARAHSIQIMKVEEIAASKCRRPAVKQFHDSKIKFPLPHRVLRRQHKPRFTTKRPNTFFSpecifications
NP_000971.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
47.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RPL18A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
6142 | |
RPL18A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKASGTLREYKVVGRCLPTPKCHTPPLYRMRIFAPNHVVAKSRFWYFVSQLKKMKKSSGEIVYCGQVFEKSPLRVKNFGIWLRYDSRSGTHNMYREYRDLTTAGAVTQCYRDMGARHRARAHSIQIMKVEEIAASKCRRPAVKQFHDSKIKFPLPHRVLRRQHKPRFTTKRPNTFF | |
RUO | |
RPL18A | |
Recombinant | |
wheat germ expression system |